EMAIL THIS PAGE TO A FRIEND
Scientific reports
Superresolution imaging with optical fluctuation using speckle patterns illumination.
PMID 26572283
Abstract
Superresolution fluorescence microscopy possesses an important role for the study of processes in biological cells with subdiffraction resolution. Recently, superresolution methods employing the emission properties of fluorophores have rapidly evolved due to their technical simplicity and direct applicability to existing microscopes. However, the application of these methods has been limited to samples labeled with fluorophores that can exhibit intrinsic emission properties at a restricted timescale, especially stochastic blinking. Here, we present a superresolution method that can be performed using general fluorophores, regardless of this intrinsic property. Utilizing speckle patterns illumination, temporal emission fluctuation of fluorophores is induced and controlled, from which a superresolution image can be obtained exploiting its statistical property. Using this method, we demonstrate, theoretically and experimentally, the capability to produce subdiffraction resolution images. A spatial resolution of 500 nm, 300 nm and 140 nm with 0.4, 0.5 and 1.4 NA objective lenses respectively was achieved in various samples with an enhancement factor of 1.6 compared to conventional fluorescence microscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252433
Rhodamine 6G, Dye content 99 %
C28H30N2O3 · HCl
R4127
Rhodamine 6G, Dye content ~95 %
C28H30N2O3 · HCl
83697
Rhodamine 6G, suitable for fluorescence, BioReagent
C28H30N2O3 · HCl