EMAIL THIS PAGE TO A FRIEND
ACS nano
All-Quantum-Dot Infrared Light-Emitting Diodes.
PMID 26575976
Abstract
Colloidal quantum dots (CQDs) are promising candidates for infrared electroluminescent devices. To date, CQD-based light-emitting diodes (LEDs) have employed a CQD emission layer sandwiched between carrier transport layers built using organic materials and inorganic oxides. Herein, we report the infrared LEDs that use quantum-tuned materials for each of the hole-transporting, the electron-transporting, and the light-emitting layers. We successfully tailor the bandgap and band position of each CQD-based component to produce electroluminescent devices that exhibit emission that we tune from 1220 to 1622 nm. Devices emitting at 1350 nm achieve peak external quantum efficiency up to 1.6% with a low turn-on voltage of 1.2 V, surpassing previously reported all-inorganic CQD LEDs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
675075
8-Mercaptooctanoic acid, 95%
C8H16O2S