EMAIL THIS PAGE TO A FRIEND
Journal of applied physics
Ferrohydrodynamic modeling of magnetic nanoparticle harmonic spectra for magnetic particle imaging.
PMID 26576063
Abstract
Magnetic Particle Imaging (MPI) is an emerging imaging technique that uses magnetic nanoparticles as tracers. In order to analyze the quality of nanoparticles developed for MPI, a Magnetic Particle Spectrometer (MPS) is often employed. In this paper, we describe results for predictions of the nanoparticle harmonic spectra obtained in a MPS using three models: the first uses the Langevin function, which does not take into account finite magnetic relaxation; the second model uses the magnetization equation by Shliomis (Sh), which takes into account finite magnetic relaxation using a constant characteristic time scale; and the third model uses the magnetization equation derived by Martsenyuk, Raikher, and Shliomis (MRSh), which takes into account the effect of magnetic field magnitude on the magnetic relaxation time. We make comparisons between these models and with experiments in order to illustrate the effects of field-dependent relaxation in the MPS. The models results suggest that finite relaxation results in a significant drop in signal intensity (magnitude of individual harmonics) and in faster spectral decay. Interestingly, when field dependence of the magnetic relaxation time was taken into account, through the MRSh model, the simulations predict a significant improvement in the performance of the nanoparticles, as compared to the performance predicted by the Sh equation. The comparison between the predictions from models and experimental measurements showed excellent qualitative as well as quantitative agreement up to the 19th harmonic using the Sh and MRSh equations, highlighting the potential of ferrohydrodynamic modeling in MPI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251682
Sodium 3-mercapto-1-propanesulfonate, technical grade, 90%
C3H7NaO3S2