EMAIL THIS PAGE TO A FRIEND
Scientific reports
Lipoteichoic acid of Streptococcus oralis Uo5: a novel biochemical structure comprising an unusual phosphorylcholine substitution pattern compared to Streptococcus pneumoniae.
PMID 26577602
Abstract
Members of the Mitis group of streptococci possess teichoic acids (TAs) as integral components of their cell wall that are unique among Gram-positive bacteria. Both, lipoteichoic (LTA) and wall teichoic acid, are formed by the same biosynthetic pathway, are of high complexity and contain phosphorylcholine (P-Cho) residues. These residues serve as anchors for choline-binding proteins (CBPs), some of which have been identified as virulence factors of the human pathogen Streptococcus pneumoniae. We investigated the LTA structure of its close relative Streptococcus oralis. Our analysis revealed that S. oralis Uo5 LTA has an overall architecture similar to pneumococcal LTA (pnLTA) and can be considered as a subtype of type IV LTA. Its structural complexity is even higher than that of pnLTA and its composition differs in number and type of carbohydrate moieties, inter-residue connectivities and especially the P-Cho substitution pattern. Here, we report the occurrence of a saccharide moiety substituted with two P-Cho residues, which is unique as yet in bacterial derived surface carbohydrates. Finally, we could link the observed important structural variations between S. oralis and S. pneumoniae LTA to the divergent enzymatic repertoire for their TA biosynthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B7906
1-Butanol, for molecular biology, ≥99%
C4H10O
281549
1-Butanol, anhydrous, 99.8%
C4H10O
154679
1-Butanol, spectrophotometric grade, 99.5%
C4H10O
270679
1-Butanol, for HPLC, 99.8%
C4H10O
442665
1-Butanol, analytical standard
C4H10O
03-4585
1-Butanol, for HPLC
C4H10O
279544
1-Propanol, anhydrous, 99.7%
C3H8O
W292818
1-Propanol, ≥99%, FG
C3H8O
W292826
1-Propanol, natural, ≥98%, FG
C3H8O
24-6080
1-Propanol, SAJ first grade, ≥99.0%
C3H8O
24-6070
1-Propanol, JIS special grade
C3H8O
A1542
Ammonium acetate, for molecular biology, ≥98%
C2H7NO2
A7330
Ammonium acetate, BioXtra, ≥98%
C2H7NO2
372331
Ammonium acetate, 99.999% trace metals basis
C2H7NO2
431311
Ammonium acetate, ≥99.99% trace metals basis
C2H7NO2
238074
Ammonium acetate, ACS reagent, ≥97%
C2H7NO2
A7262
Ammonium acetate, reagent grade, ≥98%
C2H7NO2
A2706
Ammonium acetate solution, for molecular biology, 7.5 M
C2H7NO2
W217816
Butyl alcohol, natural, ≥99.5%, FCC, FG
C4H10O
W217808
Butyl alcohol, ≥99.9%, FCC
C4H10O
633178
Deuterium oxide, 99.994 atom % D, contains 1 mM terephthalic acid disodium salt, 0.01 % (w/v) DSS-d6
D2O
613398
Deuterium oxide, extra, 99.994 atom % D
D2O
191701
Deuterium oxide, "100%", 99.990 atom % D
D2O
364312
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
D2O
151890
Deuterium oxide, "100%", ≥99.96 atom % D
D2O
269786
Deuterium oxide, "100%", 99.96 atom % D
D2O
151882
Deuterium oxide, 99.9 atom % D
D2O
530867
Deuterium oxide, 99.9 atom % D
D2O
613444
Deuterium oxide, 99.9 atom % D, glass distilled
D2O
450510
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
D2O