EMAIL THIS PAGE TO A FRIEND
Cancer research
Fibulin-5 Blocks Microenvironmental ROS in Pancreatic Cancer.
PMID 26577699
Abstract
Elevated oxidative stress is an aberration seen in many solid tumors, and exploiting this biochemical difference has the potential to enhance the efficacy of anticancer agents. Homeostasis of reactive oxygen species (ROS) is important for normal cell function, but excessive production of ROS can result in cellular toxicity, and therefore ROS levels must be balanced finely. Here, we highlight the relationship between the extracellular matrix and ROS production by reporting a novel function of the matricellular protein Fibulin-5 (Fbln5). We used genetically engineered mouse models of pancreatic ductal adenocarcinoma (PDAC) and found that mutation of the integrin-binding domain of Fbln5 led to decreased tumor growth, increased survival, and enhanced chemoresponse to standard PDAC therapies. Through mechanistic investigations, we found that improved survival was due to increased levels of oxidative stress in Fbln5-mutant tumors. Furthermore, loss of the Fbln5-integrin interaction augmented fibronectin signaling, driving integrin-induced ROS production in a 5-lipooxygenase-dependent manner. These data indicate that Fbln5 promotes PDAC progression by functioning as a molecular rheostat that modulates cell-ECM interactions to reduce ROS production, and thus tip the balance in favor of tumor cell survival and treatment-refractory disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA000868
Anti-FBLN5 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
D7008
Dihydroethidium, ≥95%
C21H21N3
37291
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
C21H21N3
M5625
Menadione, crystalline
C11H8O2
M9429
Menadione, meets USP testing specifications
C11H8O2
M57405
Menadione, 98%
C11H8O2
47775
Menadione (K3), analytical standard
C11H8O2
A7250
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
C5H9NO3S
A5099
N-Acetyl-L-cysteine, PharmaGrade, Ajinomoto, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H9NO3S
A8199
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
C5H9NO3S
A9165
N-Acetyl-L-cysteine, cell culture tested, BioReagent
C5H9NO3S
74540
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
C18H22O4
N5023
Nordihydroguaiaretic acid, ≥90% (HPLC), from Larrea divaricata (creosote bush)
C18H22O4
295647
Phosphine, electronic grade, ≥99.9995%
H3P
04004
Phosphorus, red, ≥97.0%
P
343242
Phosphorus, red, ≥99.99% trace metals basis
P
209678
Phosphorus, red, powder, 99% trace metals basis
P
R8875
Rotenone, ≥95%
C23H22O6
S1827
Sodium deoxycholate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production
C24H39NaO4