EMAIL THIS PAGE TO A FRIEND
The European physical journal. E, Soft matter
Liquid-body resonance while contacting a rotating superhydrophobic surface.
PMID 26577818
Abstract
We advance a scheme in which a liquid body on a stationary tip in contact with a rotating superhydrophobic surface is able to maintain resonance primarily from stick-slip events. With tip-to-surface spacing in the range 2.73 ≤ h < 2.45 mm for a volume of 10 μL, the liquid body was found to exhibit resonance independent of the speed of the drum. The mechanics were found to be due to a surface-tension-controlled vibration mode based on the natural frequency values determined. With spacing in the range 2.45 ≤ h < 2.15 mm imposed for a volume of 10 μL, the contact length of the liquid body was found to vary with rotation of the SH drum. This was due to the stick-slip events being able to generate higher energy fluctuations causing the liquid-solid contact areas to vary since the almost oblate spheroid shape of the liquid body had intrinsically higher surface energies. This resulted in the natural frequency perturbations being frequency- and amplitude-modulated over a lower frequency carrier. These findings have positive implications for microfluidic sensing.