EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
1,2,3-Triazole pharmacophore-based benzofused nitrogen/sulfur heterocycles with potential anti-Moraxella catarrhalis activity.
PMID 26578325
Abstract
Versatile 1,2,3-triazole pharmacophore-based benzofused heterocycles containing halogen-substituted aromatic (9-17 and 25-28), 7-substituted coumarin (18-23 and 29-30) or penciclovir-like subunit (31a,b-38a) were designed and synthesized to evaluate their antibacterial activities against selected Gram-positive and Gram-negative bacteria. Hybridization approach using environmentally friendly Cu(I)-catalyzed click reaction under microwave irradiation was adopted in the synthesis of regioselective 1,4-disubstituted 1,2,3-triazole tethered heterocycles (9-23 and 25-30), while post-N-alkylation of NH-1,2,3-triazoles afforded both 2,4- (31a-38a) and 1,4-disubstituted (31b-33b, 35b-37b) 1,2,3-triazole regioisomers. The compounds 18-23 and 25-30 revealed fluorescence in the violet region of the visible spectrum with a strong influence of phenyl spacer in 25-30 on both wavelength and emission intensity. Fusion of selected subunits led to new hybrid architecture, benzothiazole-1,2,3-triazole-coumarin 29 that demonstrated extremely narrow spectrum activity towards fastidious Gram-negative bacteria Moraxella catarrhalis. Selected hybrid showed the potency against Moraxella catarrhalis (MIC⩽0.25μg/mL) comparable to that of reference antibiotic azithromycin, which suggested that further investigations are necessary to optimize this potential hit compound as a new anti-Moraxella catarrhalis agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75199
Azithromycin, analytical standard
C38H72N2O12