EMAIL THIS PAGE TO A FRIEND
Scientific reports
Dendritic cells and macrophages neurally hard-wired in the lymph node.
PMID 26581550
Abstract
The neural hard-wired pathways in which the lymphoid organs are innervated by the nervous system is of special interest with respect to suggested afferent and sensory systems informing the central nervous system about the status of the immune system. Until today efferent also like afferent innervation seem to be unspecific, targeting many types of cells by affecting many cells at the same time. We for the first time show that antigen presenting cells (APC) are abundantly innervated in the T-cell enriched area, the subsinoidal layer and the cortical extrafollicular zone of lymph nodes in rats by a mesh of filamentous neurofilament positive structures originating from single nerve fibers and covering each single APC similar to a glass fishing float, so that we termed them "wired" APC (wAPC). These wAPC also found in humans seem to be restricted to the cell body, not to follow membranous extensions, they may be dynamic and receptive as MAP2 is expressed and axonal growth cones can be detected and they probably lack vesicular activity through missing synaptophysin expression. The specific innervation targeting single cells which show a distribution divided in several areas in one lymph node suggests a form of topographically organized afferent sensory system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503583
Anti-CD4, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1401062
Anti-CD68 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2700244
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA048982
Anti-CD68 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107746
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2107747
Anti-CD68 antibody produced in rabbit, affinity isolated antibody
SAB1400126
Anti-HLA-E antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1401182
Anti-HLA-E antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA031454
Anti-HLA-E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21349 Anti-ITGAE antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA036313
Anti-ITGAE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501861
Anti-NFASC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008832 Anti-NFASC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21487 Anti-PTPNS1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1408349
Anti-SIRPA antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2102154
Anti-SIRPA antibody produced in rabbit, affinity isolated antibody
SAB2501275
Anti-THY1 antibody produced in goat, affinity isolated antibody
HPA003733
Anti-THY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700059
Monoclonal Anti-CD4 , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
C1805 Monoclonal Anti-CD4 antibody produced in mouse, clone Q4120, purified immunoglobulin, buffered aqueous solution
SAB4700054 Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-16, purified immunoglobulin, buffered aqueous solution
SAB4700065
Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-115, purified immunoglobulin, buffered aqueous solution
SAB4700733 Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
SAB4100489
Monoclonal Anti-CD4 antibody produced in mouse, clone CD4 -A409, culture supernatant
AMAB90754
Monoclonal Anti-CD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0395, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90873
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1338, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90874
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1346, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700300 Monoclonal Anti-HLA-E antibody produced in mouse, clone MEM-E/08, purified immunoglobulin, buffered aqueous solution