EMAIL THIS PAGE TO A FRIEND
Glycobiology
Schistosoma mansoni α1,3-fucosyltransferase-F generates the Lewis X antigen.
PMID 26582608
Abstract
Genetic evidence suggests that the Schistosoma mansoni genome contains six genes that encode α1,3-fucosyltransferases (smFuTs). To date, the activities and specificities of these putative fucosyltransferases are unknown. As Schistosoma express a variety of fucosylated glycans, including the Lewis X antigen Galβ1-4(Fucα1-3)GlcNAcβ-R, it is likely that this family of genes encode enzymes that are partly responsible for the generation of those structures. Here, we report the molecular cloning of fucosyltransferase-F (smFuT-F) from S. mansoni, as a soluble, green fluorescent protein fusion protein and its acceptor specificity. The gene smFuT-F was expressed in HEK freestyle cells, purified by affinity chromatography, and analyzed toward a broad panel of glycan acceptors. The enzyme product of smFuT-F effectively utilizes a type II chain acceptor Galβ1-4GlcNAc-R, but notably not the LDN sequence GalNAcβ1-4GlcNAc-R, to generate Lewis X type-glycans, and smFuT-F transcripts are present in all intramammalian life stages.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4401
Guanosine 5′-diphospho-β-L-fucose sodium salt, ≥85%
C16H23N5Na2O15P2