EMAIL THIS PAGE TO A FRIEND
Journal of dairy science
Short communication: Drug residues in goat milk after prophylactic use of antibiotics in intravaginal sponges for estrus synchronization.
PMID 26585470
Abstract
The aim of this study was to determine whether the prophylactic use of antibiotics in intravaginal sponges used for estrus synchronization in goats may result in the presence of inhibitors in milk and, therefore, of positive results by microbial screening tests. Ninety-eight Murciano-Granadina goats were used, divided into 7 groups of 14 animals. Intravaginal sponges were placed in 6 groups using 2 concentrations of 3 different antibiotics: doxycycline, oxytetracycline, and sulfathiazole-framycetin. The sponges of the control group were placed without antibiotics. Milk samples were collected daily until 7 d posttreatment and analyzed using 3 microbial tests. Positive samples were retested by specific receptor-binding assays to confirm the positive results. Vaginal status was evaluated by visual assessment of the external aspect of the sponges after removal. The microbial test response was not affected by either day posttreatment or dose of antibiotic used, except for oxytetracycline at the higher concentration. Moreover, no positive results were obtained using receptor-binding assays, suggesting that residues, if present in milk, did not exceed the regulatory (safety) levels established for these drugs. The occurrence of soiled sponges was higher in the control group. With respect to the dose of antibiotics used, no significant differences were found for the lower dose administered. However, a significant increase in the percentage of clean sponges was observed for the higher dose of doxycycline. We conclude that the prophylactic use of low doses of doxycycline, oxytetracycline, or sulfathiazole in intravaginal sponges used for synchronization of estrus helps to reduce clinical vaginitis in dairy goats and does not seem to be the cause of positive results in microbial inhibitor tests used to detect antibiotics in goat milk.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N3398
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
C7H12N2O4S