EMAIL THIS PAGE TO A FRIEND
Genome biology
Accelerating research through reagent repositories: the genome editing example.
PMID 26585970
Abstract
Keith Joung, Dan Voytas and Joanne Kamens share insights into how the genome editing field was advanced by early access to biological resources and the role in this process that plasmid repositories play.