EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Two bisthienylethene-Ir(III) complexes showing acid/base-induced structural transformation and on-off luminescence switching in solution.
PMID 26586364
Abstract
Complexes [Ir(dfppy)2(pbdtiH)](PF6)·2CHCl3 (1-H) and [Ir(dfppy)2(pbdti)] (1) were synthesized by the reaction of bisthienylethene pbdtiH and an [Ir(dfppy)2Cl]2 dimer under neutral and basic conditions, respectively. Thus, the {Ir(dfppy)2}(+) unit is coordinated by pbdtiH in 1-H, and by pbdti(-) in 1, which are confirmed by their crystal structures. The structures of 1-H and 1 could be interconverted in solution, upon alternately adding NEt3 and TFA, thus resulting in reversible luminescence switching between the on-state of 1-H and the off-state of 1 at room temperature. In addition, both 1-H and 1 show solid-state luminescence, with a broad emission at 534 nm and 525 nm, respectively. The free pbdtiH ligand shows photochromic behavior in CH2Cl2 solution. However, no photochromism has been observed in 1-H and 1, indicating that the coordination of the pbdtiH/pbdti(-) ligand to the {Ir(dfppy)2}(+) unit could suppress their photochromic behaviors.