EMAIL THIS PAGE TO A FRIEND
Cytotechnology
Mitigating effect of Indian propolis against mitomycin C induced bone marrow toxicity.
PMID 26590833
Abstract
A major drawback with cancer chemotherapy is its severe toxic effects on non-target tissues. Assessment of natural products for their protective effect against anticancer drugs-induced toxicity is gaining importance in cancer biology. The present study was aimed at assessing the protective effect of hydroethanolic extract of Indian propolis (HEIP) against mitomycin C (MMC)-induced genotoxicity and cytotoxicity. Swiss albino mice were injected with various doses of HEIP (100, 200, 300, 400, 600 and 800xa0mg/kg b. wt., i.p) 1xa0h prior to MMC (8xa0mg/kg, i.p.) injection. The geno- and cyto-toxicities were evaluated in mice by performing bone marrow micronucleus and TUNEL assays. In vitro antioxidant and lipid peroxidation inhibitory assays were carried out to understand the mechanism of the protective effects. The significant increase in the frequency of micronculeated cells (12.51xa0±xa00.48), apoptotic cells (23.43xa0±xa01.86) and reduction in P/N ratio (0.69xa0±xa00.04) compared with control indicated the potential geno- and cytotoxic effects of MMC in bone marrow. Pretreatment with HEIP resulted in the significant recovery of the toxic effects induced by MMC. HEIP at 400xa0mg/kgxa0b. wt. was found to be the optimum dose imparting the maximum protective effects. The in vitro antioxidant and lipid peroxidation inhibitory assays suggest that the extract possesses substantial free radical scavenging activities. In conclusion, HEIP possesses substantial geno- and cyto-protective properties against MMC, which could be mediated through efficient free radical scavenging and inhibitory effect on lipid peroxidation.