EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Fabrication and characterization of a highly sensitive hydroquinone chemical sensor based on iron-doped ZnO nanorods.
PMID 26592548
Abstract
Herein, we report the development of a simple and highly sensitive hydroquinone (HQ) chemical sensor based on an electrochemically activated iron-doped (Fe-doped) zinc oxide nanorod (ZnO NR) modified screen-printed electrode (SPE). The Fe-doped ZnO NRs were prepared using a hydrothermal process and their morphological, crystal, compositional and optical properties were characterized in detail. The detailed characterizations showed that the NRs are densely grown, well-crystalline and possess a wurtzite hexagonal phase. The fabricated HQ electrochemical sensor exhibited high sensitivity of 18.60 μA mM(-1) cm(-2) and a very low experimental detection limit of 0.51 μM. Our results demonstrate that simply prepared doped ZnO nanomaterials are promising candidates to fabricate highly sensitive electrochemical sensors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
338818
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
H5NO
320145
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
H5NO
221228
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
H5NO
17093
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
H5NO
30501
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
H5NO
05002
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
H5NO
05003
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
H5NO
40205
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)
H5NO
40291
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)
H5NO
40321
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade SLSI PURANAL (Honeywell 17301)
H5NO
01-4362
Ammonium hydroxide solution, 28% NH3 in H2O, suitable for determination of toxic metals
H5NO
H6404
Hexamethylenetetramine, BioXtra
C6H12N4
33233
Hexamethylenetetramine, puriss. p.a., Reag. Ph. Eur., ≥99.5% (calc. to the dried substance)
C6H12N4
15614
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
C6H12N4
398160
Hexamethylenetetramine, ACS reagent, ≥99.0%
C6H12N4
H11300
Hexamethylenetetramine, ReagentPlus®, 99%
C6H12N4
H9003
Hydroquinone, ReagentPlus®, ≥99%
C6H6O2
H3660
Hydroquinone, meets USP testing specifications
C6H6O2
H17902
Hydroquinone, ReagentPlus®, ≥99.5%
C6H6O2