EMAIL THIS PAGE TO A FRIEND
Talanta
Rapid assessment of bioactive phenolics and methylxanthines in spent coffee grounds by FT-NIR spectroscopy.
PMID 26592633
Abstract
Spent coffee grounds (SCGs) are a great source of bioactive compounds with interest to pharmaceutical and cosmetic industries. Phenolics and methylxanthines are the main health related compounds present in SCG samples. Content estimation of these compounds in SCGs is of upmost importance in what concerns their profitable use by waste recovery industries. In the present work, near-infrared spectroscopy (NIRS) was proposed as a rapid and non-destructive technique to assess the content of three main phenolics (caffeic acid, (+)-catechin and chlorogenic acid) and three methylxanthines (caffeine, theobromine and theophylline) in SCG samples obtained from different coffee brands and diverse coffee machines. The content of these compounds was determined for 61 SCG samples by HPLC coupled with diode-array detection. Partial least squares (PLS) regression based models were calibrated to correlate diffuse reflectance NIR spectra against the reference data for the six parameters obtained by HPLC. Spectral wavelength selection and number of latent variables were optimized by minimizing the cross-validation error. PLS models showed good linearity with a coefficient of determination for the prediction set (Rp(2)) of 0.95, 0.92, 0.88, 071 and 0.84 for caffeine, caffeic acid, (+)-catechin, chlorogenic acid and theophylline, respectively. The range error ratio (RER) was higher for caffeine (17.8) when compared to other compounds (12.0, 10.1, 7.6 and 9.2, respectively for caffeic acid, (+)-catechin, chlorogenic acid and theophylline). Moreover, the content of caffeine could be used to predict the antioxidant properties of SCG samples (R=0.808, n=61), despite not presenting this property itself. The results obtained confirmed that NIRS is a suitable technique to screen SCG samples unveiling those with high content of bioactive compounds, which are interesting for subsequent extraction procedures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8380
5α-Androstan-17β-ol-3-one, ≥97.5%
C19H30O2
45726
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
C2H4O2
W200603
Acetic acid, natural, ≥99.5%, FG
C2H4O2
W200611
Acetic acid, ≥99.5%, FCC, FG
C2H4O2
ARK2183
Acetic acid, glacial, PharmaGrade, USP, JP, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C2H4O2
606766
Acetic acid-12C2, 99.9 atom % 12C
12C2H4O2
C0625
Caffeic acid, ≥98.0% (HPLC)
C9H8O4
60020
Caffeic acid, purum, ≥95.0% (HPLC)
C9H8O4
93784
Caffeine, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C8H10N4O2
C0750
Caffeine, powder, ReagentPlus®
C8H10N4O2
C8960
Caffeine, BioXtra
C8H10N4O2
C1778
Caffeine, Sigma Reference Standard, vial of 250 mg
C8H10N4O2
C7731
Caffeine, meets USP testing specifications, anhydrous
C8H10N4O2
W222402
Caffeine, anhydrous, 99%, FCC, FG
C8H10N4O2
05-0370
Caffeine, SAJ special grade, ≥98.5%
C8H10N4O2
C3878
Chlorogenic acid, ≥95% (titration)
C16H18O9
A9967
Glacial acetic acid, meets USP testing specifications
C2H4O2
322415
Methanol, anhydrous, 99.8%
CH4O
M1770
Methanol, BioReagent, suitable for protein sequencing
CH4O
650609
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
CH4O
270474
Methanol, for HPLC, gradient grade, 99.93%
CH4O
65548
Methanol, for HPLC, gradient grade, ≥99.8% (GC)
CH4O
40217
Methanol, semiconductor grade PURANAL (Honeywell 17824)
CH4O
19-2470
Methanol, for HPLC
CH4O
19-2453
Methanol, for residue analysis, suitable for 5000 per JIS
CH4O
19-2400
Methanol, SAJ special grade
CH4O
19-2425
Methanol, suitable for determination of dioxins
CH4O
717975
Methanol, NMR reference standard
CH4O
650188
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
CH4O
632546
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, ≥99.5%
CH4O