EMAIL THIS PAGE TO A FRIEND
Scientific reports
Plant phenolics are detoxified by prophenoloxidase in the insect gut.
PMID 26592948
Abstract
Plant phenolics are a group of important secondary metabolites that are toxic to many animals and insects if ingested at high concentrations. Because most insects consume plant phenolics daily, they have likely evolved the capacity to detoxify these compounds. Here, we used Drosophila melanogaster, Bombyx mori and Helicoverpa armigera as models to study the metabolism of plant phenolics by prophenoloxidases. We found that insect foreguts release prophenoloxidases into the lumen, and that the survival of prophenoloxidase-deletion mutants was impaired when fed several plant phenolics and tea extracts. Using l-DOPA as a model substrate, biochemical assays in large Lepidopteran insects demonstrated that low levels of l-DOPA are rapidly metabolized into intermediates by phenoloxidases. Feeding with excess l-DOPA showed that the metabolic intermediate 5,6-dihydroxyindole reached the hindgut either by passing directly through the midgut, or by transport through the hemolymph. In the hindgut, 5,6-dihydroxyindole was further oxidized by prophenoloxidases. Intermediates exerted no toxicity in the hemocoel or midgut. These results show that plant phenolics are not toxic to insects unless prophenoloxidase genes are lost or the levels of phenolics exceed the catalytic activity of the gut prophenoloxidases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4630
2,5-Diphenyloxazole, suitable for liquid scintillation spectrometry
C15H11NO
D210404
2,5-Diphenyloxazole, 99%, suitable for scintillation
C15H11NO
D9628
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
C9H11NO4
2858
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
C2H6O
48075
Ethanol solution, certified reference material, 2000 μg/mL in methanol
C2H6O
E2385
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
C2H6O
792780
Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792799
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
459836
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
C2H6O
792802
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
C2H6O
792829
Ethyl alcohol, Pure, 140 proof, meets water USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792810
Ethyl alcohol, Pure, 140 proof, Excise Tax-free, Permit for use required
C2H6O
493538
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
C2H6O
493511
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
C2H6O
16201
Tannic acid, puriss., meets analytical specification of USP, powder
C76H52O46
403040
Tannic acid, ACS reagent
C76H52O46
T0200
Tannic acid, Source: Chinese natural gall nuts
C76H52O46
W304204
Tannic acid
C76H52O46