EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
PAK4 promotes kinase-independent stabilization of RhoU to modulate cell adhesion.
PMID 26598620
Abstract
P21-activated kinase 4 (PAK4) is a Cdc42 effector protein thought to regulate cell adhesion disassembly in a kinase-dependent manner. We found that PAK4 expression is significantly higher in high-grade human breast cancer patient samples, whereas depletion of PAK4 modifies cell adhesion dynamics of breast cancer cells. Surprisingly, systematic analysis of PAK4 functionality revealed that PAK4-driven adhesion turnover is neither dependent on Cdc42 binding nor kinase activity. Rather, reduced expression of PAK4 leads to a concomitant loss of RhoU expression. We report that RhoU is targeted for ubiquitination by the Rab40A-Cullin 5 complex and demonstrate that PAK4 protects RhoU from ubiquitination in a kinase-independent manner. Overexpression of RhoU rescues the PAK4 depletion phenotype, whereas loss of RhoU expression reduces cell adhesion turnover and migration. These data support a new kinase-independent mechanism for PAK4 function, where an important role of PAK4 in cellular adhesions is to stabilize RhoU protein levels. Thus, PAK4 and RhoU cooperate to drive adhesion turnover and promote cell migration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300427
Anti-PAK1 (Ab-212) antibody produced in rabbit, affinity isolated antibody
SAB2500741
Anti-PAK1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406219
Anti-PAK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003565
Anti-PAK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502062
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502064
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502065
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502066
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502063
Anti-PAK1, C-Terminal antibody produced in rabbit, affinity isolated antibody
PRS3077
Anti-PAK4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2106811
Anti-PAK4 antibody produced in rabbit, affinity isolated antibody
SAB4500076
Anti-PAK4, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406298
Anti-PKN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003982
Anti-PKN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411085
Anti-PKN1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
EMU008821 MISSION® esiRNA, esiRNA targeting mouse Pak4 (esiRNA1)
EMU039321 MISSION® esiRNA, esiRNA targeting mouse Pak2 (esiRNA1)
EHU026721 MISSION® esiRNA, esiRNA human PAK2 (esiRNA1)
EHU124131 MISSION® esiRNA, esiRNA human PKN1 (esiRNA1)
EHU085541 MISSION® esiRNA, esiRNA human PKN2 (esiRNA1)
EHU073951 MISSION® esiRNA, esiRNA human PAK4 (esiRNA1)
EHU140181 MISSION® esiRNA, esiRNA human PAK1 (esiRNA1)
EMU088481 MISSION® esiRNA, esiRNA targeting mouse Stk3 (esiRNA1)
EMU026031 MISSION® esiRNA, esiRNA targeting mouse Pak1 (esiRNA1)
WH0005058M1
Monoclonal Anti-PAK1 antibody produced in mouse, clone 1E11, purified immunoglobulin, buffered aqueous solution
SAB1400193
Monoclonal Anti-PAK1 antibody produced in mouse, clone 4D1, purified immunoglobulin, buffered aqueous solution
WH0010298M1
Monoclonal Anti-PAK4 antibody produced in mouse, clone 3F10, purified immunoglobulin, buffered aqueous solution
WH0005585M1
Monoclonal Anti-PKN1 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1404244
Monoclonal Anti-PKN1 antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution