PloS one
Kaempferol Inhibits Endoplasmic Reticulum Stress-Associated Mucus Hypersecretion in Airway Epithelial Cells And Ovalbumin-Sensitized Mice.
PMID 26599511
Abstract
Mucus hypersecretion is an important pathological feature of chronic airway diseases, such as asthma and pulmonary diseases. MUC5AC is a major component of the mucus matrix forming family of mucins in the airways. The initiation of endoplasmic reticulum (ER)-mediated stress responses contributes to the pathogenesis of airway diseases. The present study investigated that ER stress was responsible for airway mucus production and this effect was blocked by the flavonoid kaempferol. Oral administration of ≥10 mg/kg kaempferol suppressed mucus secretion and goblet cell hyperplasia observed in the bronchial airway and lung of BALB/c mice sensitized with ovalbumin (OVA). TGF-β and tunicamycin promoted MUC5AC induction after 72 h in human bronchial airway epithelial BEAS-2B cells, which was dampened by 20 μM kaempferol. Kaempferol inhibited tunicamycin-induced ER stress of airway epithelial cells through disturbing the activation of the ER transmembrane sensor ATF6 and IRE1α. Additionally, this compound demoted the induction of ER chaperones such as GRP78 and HSP70 and the splicing of XBP-1 mRNA by tunicamycin. The in vivo study further revealed that kaempferol attenuated the induction of XBP-1 and IRE1α in epithelial tissues of OVA-challenged mice. TGF-β and tunicamycin induced TRAF2 with JNK activation and such induction was deterred by kaempferol. The inhibition of JNK activation encumbered the XBP-1 mRNA splicing and MUC5AC induction by tunicamycin and TGF-β. These results demonstrate that kaempferol alleviated asthmatic mucus hypersecretion through blocking bronchial epithelial ER stress via the inhibition of IRE1α-TRAF2-JNK activation. Therefore, kaempferol may be a potential therapeutic agent targeting mucus hypersecretion-associated pulmonary diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6435
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
AlKO8S2 · 12H2O
31242
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
AlKO8S2 · 12H2O
12623
Aluminum potassium sulfate dodecahydrate, puriss., meets analytical specification of Ph. Eur., BP, 99.0-100.5% (calc. on dry substance)
AlKO8S2 · 12H2O
237086
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
AlKO8S2 · 12H2O
A7210
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
AlKO8S2 · 12H2O
SAB3701027 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701127 Anti-Mouse IgG (H+L), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701157 Anti-Mouse IgG (H+L), F(ab) fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700991 Anti-Mouse IgG F(ab′)2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701013 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700849 Anti-Rabbit IgG (Fc specific), highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700946 Anti-Rabbit IgG (H+L), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700970 Anti-Rabbit IgG (H+L), F(ab) fragment antibody produced in goat, affinity isolated antibody, buffered aqueous solution
R9130 Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700840
Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
C106
CGS-12066 maleate salt, solid
C17H17F3N4 · C4H4O4
H4001
Hydrocortisone, ≥98% (HPLC)
C21H30O5
H0135
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
C21H30O5
H0888
Hydrocortisone, BioReagent, suitable for cell culture
C21H30O5
H3160
Hydrocortisone, meets USP testing specifications
C21H30O5
P7971
Potassium alum dodecahydrate, meets USP testing specifications
AlKO8S2 · 12H2O