EMAIL THIS PAGE TO A FRIEND
Scientific reports
RNA-based, transient modulation of gene expression in human haematopoietic stem and progenitor cells.
PMID 26599627
Abstract
Modulation of gene expression is a useful tool to study the biology of haematopoietic stem and progenitor cells (HSPCs) and might also be instrumental to expand these cells for therapeutic approaches. Most of the studies so far have employed stable gene modification by viral vectors that are burdensome when translating protocols into clinical settings. Our study aimed at exploring new ways to transiently modify HSPC gene expression using non-integrating, RNA-based molecules. First, we tested different methods to deliver these molecules into HSPCs. The delivery of siRNAs with chemical transfection methods such as lipofection or cationic polymers did not lead to target knockdown, although we observed more than 90% fluorescent cells using a fluorochrome-coupled siRNA. Confocal microscopic analysis revealed that despite extensive washing, siRNA stuck to or in the cell surface, thereby mimicking a transfection event. In contrast, electroporation resulted in efficient, siRNA-mediated protein knockdown. For transient overexpression of proteins, we used optimised mRNA molecules with modified 5'- and 3'-UTRs. Electroporation of mRNA encoding GFP resulted in fast, efficient and persistent protein expression for at least seven days. Our data provide a broad-ranging comparison of transfection methods for hard-to-transfect cells and offer new opportunities for DNA-free, non-integrating gene modulation in HSPCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74875
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
C24H21Na3O11S3
01810
Cycloheximide, ≥93.0% (HPLC)
C15H23NO4
C7698
Cycloheximide, from microbial, ≥94% (TLC)
C15H23NO4
C1988
Cycloheximide, Biotechnology Performance Certified
C15H23NO4
C4859
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, 0.2 μm filtered
C15H23NO4
D2650
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
C2H6OS
D8418
Dimethyl sulfoxide, for molecular biology
C2H6OS
D4540
Dimethyl sulfoxide, ≥99.5% (GC), plant cell culture tested
C2H6OS
D1435
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
C2H6OS
D9170
Dimethyl sulfoxide, PCR Reagent
C2H6OS
D2438
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
C2H6OS
276855
Dimethyl sulfoxide, anhydrous, ≥99.9%
C2H6OS
41639
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
C2H6OS
W387509
Dimethyl sulfoxide, ≥99%
C2H6OS
07-4860
Dimethyl sulfoxide, JIS special grade, ≥99.0%
C2H6OS
07-4871
Dimethyl sulfoxide, for HPLC
C2H6OS
673439 Dimethyl sulfoxide solution, 50 wt. % in H2O
C2H6OS
606987
Dimethyl sulfoxide-12C2, 99.95 atom % 12C
C2H6SO
46955
Fluorescein, for fluorescence, free acid
C20H12O5
EHU001391 MISSION® esiRNA, esiRNA targeting human TMEM131 (esiRNA1)
EHU011481 MISSION® esiRNA, esiRNA targeting human PROM1 (esiRNA1)
EMU055321 MISSION® esiRNA, esiRNA targeting mouse Prom1 (esiRNA1)
EMU062291 MISSION® esiRNA, esiRNA targeting mouse Tmem131 (esiRNA1)
P6061
Phenylacetic acid, plant cell culture tested
C8H8O2
P16621
Phenylacetic acid, 99%
C8H8O2
W287814
Phenylacetic acid, natural, ≥99%
C8H8O2
W287806
Phenylacetic acid, ≥99%, FCC, FG
C8H8O2