EMAIL THIS PAGE TO A FRIEND
Nature
Immune homeostasis enforced by co-localized effector and regulatory T cells.
PMID 26605524
Abstract
FOXP3(+) regulatory T cells (Treg cells) prevent autoimmunity by limiting the effector activity of T cells that have escaped thymic negative selection or peripheral inactivation. Despite the information available about molecular factors mediating the suppressive function of Treg cells, the relevant cellular events in intact tissues remain largely unexplored, and whether Treg cells prevent activation of self-specific T cells or primarily limit damage from such cells has not been determined. Here we use multiplex, quantitative imaging in mice to show that, within secondary lymphoid tissues, highly suppressive Treg cells expressing phosphorylated STAT5 exist in discrete clusters with rare IL-2-positive T cells that are activated by self-antigens. This local IL-2 induction of STAT5 phosphorylation in Treg cells is part of a feedback circuit that limits further autoimmune responses. Inducible ablation of T cell receptor expression by Treg cells reduces their regulatory capacity and disrupts their localization in clusters, resulting in uncontrolled effector T cell responses. Our data thus reveal that autoreactive T cells are activated to cytokine production on a regular basis, with physically co-clustering T cell receptor-stimulated Treg cells responding in a negative feedback manner to suppress incipient autoimmunity and maintain immune homeostasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2700362
Anti-Foxp3 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW22461F Anti-Interleukin-2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
I8398 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I8273 Anti-Interleukin-2 antibody produced in goat, IgG fraction of antiserum, lyophilized powder
I7401 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
I7784 Anti-Interleukin-2 antibody produced in goat, affinity isolated antibody, lyophilized powder
HPA017357
Anti-NT5E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0003558M3 Monoclonal Anti-IL2 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution
SAB1403970
Monoclonal Anti-IL2, (C-terminal) antibody produced in mouse, clone 1C5, purified immunoglobulin, buffered aqueous solution
I6784 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5334, purified immunoglobulin, lyophilized powder
I6909 Monoclonal Anti-Interleukin-2 antibody produced in mouse, clone 5355, purified immunoglobulin, lyophilized powder
WH0004907M1
Monoclonal Anti-NT5E antibody produced in mouse, clone 4C4-2B5, purified immunoglobulin, buffered aqueous solution
SAB5300408 Monoclonal Anti-NT5E antibody produced in mouse, clone 1D7, ascites fluid
SAB1404149
Monoclonal Anti-NT5E, (C-terminal) antibody produced in mouse, clone 4D3-2C10, purified immunoglobulin, buffered aqueous solution
SAB1402533
Monoclonal Anti-NTE, (C-terminal) antibody produced in mouse, clone 3D10, purified immunoglobulin, buffered aqueous solution