PloS one
An Atherogenic Paigen-Diet Aggravates Nephropathy in Type 2 Diabetic OLETF Rats.
PMID 26606054
Abstract
Diabetic nephropathy develops in association with hyperglycemia, is aggravated by atherogenic factors such as dyslipidemia, and is sometimes initiated before obvious hyperglycemia is seen. However, the precise mechanisms of progression are still unclear. In this study, we investigated the influence of an atherogenic Paigen diet (PD) on the progression of nephropathy in spontaneous type 2 diabetic OLETF rats. Feeding PD to male OLETF rats for 12 weeks caused an extensive increase in excretion of urinary albumin and markers of tubular injury such as KIM-1 and L-FABP, accompanied by mesangial expansion and tubular atrophy. PD significantly increased plasma total cholesterol concentration, which correlates well with increases in urine albumin excretion and mesangial expansion. Conversely, PD did not change plasma glucose and free fatty acid concentrations. PD enhanced renal levels of mRNA for inflammatory molecules such as KIM-1, MCP-1, TLR4 and TNF-α and promoted macrophage infiltration and lipid accumulation in the tubulointerstitium and glomeruli in OLETF rats. Intriguingly, PD had little effect on urine albumin excretion and renal morphology in normal control LETO rats. This model may be useful in studying the complex mechanisms that aggravate diabetic nephropathy in an atherogenic environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H5653
8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)
C10H13N5O5
SAB1401062
Anti-CD68 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2700244
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA048982
Anti-CD68 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107746
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2107747
Anti-CD68 antibody produced in rabbit, affinity isolated antibody
C75209
Cholesterol, 94%
C27H46O
C8667
Cholesterol, Sigma Grade, ≥99%
C27H46O
C8503
Cholesterol, from sheep wool, ≥92.5% (GC), powder
C27H46O
C3045
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
C27H46O
C3292
Cholesterol, ≥95% (GC), powder, Ash, free
C27H46O
H6167
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
C27H46O
47127-U
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
C27H46O
C1231
Cholesterol, Plant-Derived, SyntheChol®, PharmaGrade, USP/NF, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C27H46O
AMAB90873
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1338, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90874
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1346, purified immunoglobulin, buffered aqueous glycerol solution
S5442
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
C27H46O