EMAIL THIS PAGE TO A FRIEND
Scientific reports
In-depth proteomic analysis of shell matrix proteins of Pinctada fucata.
PMID 26608573
Abstract
The shells of pearl oysters, Pinctada fucata, are composed of calcite and aragonite and possess remarkable mechanical properties. These shells are formed under the regulation of macromolecules, especially shell matrix proteins (SMPs). Identification of diverse SMPs will lay a foundation for understanding biomineralization process. Here, we identified 72 unique SMPs using liquid chromatography-tandem mass spectrometry (LC-MS/MS) analysis of proteins extracted from the shells of P. fucata combined with a draft genome. Of 72 SMPs, 17 SMPs are related to both the prismatic and nacreous layers. Moreover, according to the diverse domains found in the SMPs, we hypothesize that in addition to controlling CaCO3 crystallization and crystal organization, these proteins may potentially regulate the extracellular microenvironment and communicate between cells and the extracellular matrix (ECM). Immunohistological localization techniques identify the SMPs in the mantle, shells and synthetic calcite. Together, these proteomic data increase the repertoires of the shell matrix proteins in P. fucata and suggest that shell formation in P. fucata may involve tight regulation of cellular activities and the extracellular microenvironment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
271004
Acetonitrile, anhydrous, 99.8%
C2H3N
675415
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
C2H3N
437557
Acetonitrile, ≥99.5%, ACS reagent
C2H3N
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, suitable for 5000 per JIS
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
639133
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
D8284
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
C20H10N2Na2O4 · xH2O
74255
Sodium dodecyl sulfate, dust-free pellets, ≥99.0% (GC)
C12H25NaO4S
L3771
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
C12H25NaO4S
L6026
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
C12H25NaO4S
436143
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
C12H25NaO4S
L4509
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
C12H25NaO4S
L5750
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
C12H25NaO4S
71725
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
71729
Sodium dodecyl sulfate, ≥98.0% (GC)
C12H25NaO4S
71717
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
C12H25NaO4S
62862
Sodium dodecyl sulfate, ≥90%
C12H25NaO4S
75746
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
C12H25NaO4S