EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Use of PERCIST for Prediction of Progression-Free and Overall Survival After Radioembolization for Liver Metastases from Pancreatic Cancer.
PMID 26609176
Abstract
We evaluated the prognostic accuracy of established PET response criteria in patients with liver metastases from pancreatic cancer after treatment with (90)Y microspheres. Seventeen patients underwent (18)F-FDG PET/CT before and 3 mo after radioembolization for liver metastases from pancreatic cancer. Overall survival, progression-free survival, and time to intrahepatic progression were among other factors correlated with metabolic response as revealed by PERCIST 1.0-defined declining SUVpeak and total-lesion glycolysis. Metabolic response by change in SUVpeak (7/17) and change in total-lesion glycolysis (7/17) was a predictor for overall survival (P = 0.039; hazard ratio [HR], 0.24; 95% confidence interval [CI], 0.06-0.93), progression-free survival (P = 0.016; HR, 0.15; 95% CI, 0.03-0.69), and time to intrahepatic progression (P = 0.010; HR, 0.16; 95% CI, 0.04-0.65). A summed baseline CT diameter of less than 8 cm for the 2 largest liver metastases predicted time to intrahepatic progression (P = 0.013; HR, 0.21; 95% CI, 0.06-0.72) but did not predict overall or progression-free survival. Patient outcome was not predicted by other parameters, including baseline SUVpeak, baseline total-lesion glycolysis, or change in serum level of carcinoembryonic antigen or carbohydrate antigen 19-9 from baseline to follow-up (each, P > 0.05). Metabolic response by (18)F-FDG PET/CT predicts overall survival, progression-free survival, and time to intrahepatic progression after radioembolization for liver metastases from pancreatic cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F2756
Fluorescein di(β-D-galactopyranoside)
C32H32O15