EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
IL-10 derived from CD1dhiCD5⁺ B cells regulates experimental autoimmune myasthenia gravis.
PMID 26616882
Abstract
IL-10-competent subset within CD1d(hi)CD5(+) B cells, also known as B10 cells, has been shown to regulate autoimmune diseases. In our previous study, adoptive transfer of CD1d(hi)CD5(+) B cells expanded in vivo by GM-CSF prevented and suppressed experimental autoimmune myasthenia gravis (EAMG). The goal of this study was to further examine the role and mechanism of IL-10 in the regulatory function of B10 cells in EAMG. We found that only IL-10 competent CD1d(hi)CD5(+) B cells sorted from WT mice, but not IL-10 deficient CD1d(hi)CD5(+) B cells exhibited regulatory function in vitro and in vivo. Adoptive transfer of IL-10 competent CD1d(hi)CD5(+) B cells led to higher frequency of Tregs and B10 cells, and low levels of proinflammatory cytokines and autoantibody production. We conclude that IL-10 production within CD1d(hi)CD5(+) B cells plays an important role in immune regulation of EAMG.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0804 Mouse IgG2b ELISA Kit