EMAIL THIS PAGE TO A FRIEND
Scientific reports
RBFOX3/NeuN is Required for Hippocampal Circuit Balance and Function.
PMID 26619789
Abstract
RBFOX3 mutations are linked to epilepsy and cognitive impairments, but the underlying pathophysiology of these disorders is poorly understood. Here we report replication of human symptoms in a mouse model with disrupted Rbfox3. Rbfox3 knockout mice displayed increased seizure susceptibility and decreased anxiety-related behaviors. Focusing on hippocampal phenotypes, we found Rbfox3 knockout mice showed increased expression of plasticity genes Egr4 and Arc, and the synaptic transmission and plasticity were defective in the mutant perforant pathway. The mutant dentate granules cells exhibited an increased frequency, but normal amplitude, of excitatory synaptic events, and this change was associated with an increase in the neurotransmitter release probability and dendritic spine density. Together, our results demonstrate anatomical and functional abnormality in Rbfox3 knockout mice, and may provide mechanistic insights for RBFOX3-related human brain disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K3375
Kynurenic acid, ≥98%
C10H7NO3
S0532
Strychnine
C21H22N2O2