EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Camellia sinensis Ameliorates the Efficacy of Last Line Antibiotics Against Carbapenem Resistant Escherichia coli.
PMID 26620305
Abstract
Aquo-ethanolic extract of Camellia sinensis (PTRC-31911-A), standardized using Fourier transform infrared analysis, was found to have seven common functional groups in comparison with pre-identified marker compound 'quercetin'. Phyto-chemical quantitation analysis revealed the presence of 10.65 µg/mg of flavonoids. The bioactivity fingerprint profile of PTRC-31911-A includes IC50 (Hydroxyl radical site specific scavenging)  = 11.36 ± 0.5 µg/mL, IC80 (Hydroxyl radical non-site specific scavenging)  = 26.44 ± 0.5 µg/mL and IC50 (Superoxide ion scavenging)  = 10.141 ± 0.5 µg/mL. The drug combination analysis of PTRC-31911-A with five third-line antibiotics was carried out against carbapenem-resistant Escherichia coli. The analysis of combination of PTRC-31911-A (6.25-1000 µg/mL) and antibiotics (6.25-1000 µg/mL) revealed synergistic behaviour (fractional inhibitory concentration indices < 1) with tigecycline, ertapenem, meropenem, colistin and augmentin. The lead combination of PTRC-31911-A + ertapenem or meropenem showed maximum augmentative potential at 50 and 100 µg/mL, respectively, with nearly five-fold decrease in minimum inhibitory concentrations as compared with respective antibiotics alone. The synergistic effects implied that the antibacterial combinations of PTRC-31911-A and ertapenem, meropenem, colistin, tigecycline or augmentin would be more effective than a single monotherapy with either of the antibacterial agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS022172
Ertapenem
C22H25N3O7S