EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
An Asp-CaM complex is required for centrosome-pole cohesion and centrosome inheritance in neural stem cells.
PMID 26620907
Abstract
The interaction between centrosomes and mitotic spindle poles is important for efficient spindle formation, orientation, and cell polarity. However, our understanding of the dynamics of this relationship and implications for tissue homeostasis remains poorly understood. Here we report that Drosophila melanogaster calmodulin (CaM) regulates the ability of the microcephaly-associated protein, abnormal spindle (Asp), to cross-link spindle microtubules. Both proteins colocalize on spindles and move toward spindle poles, suggesting that they form a complex. Our binding and structure-function analysis support this hypothesis. Disruption of the Asp-CaM interaction alone leads to unfocused spindle poles and centrosome detachment. This behavior leads to randomly inherited centrosomes after neuroblast division. We further show that spindle polarity is maintained in neuroblasts despite centrosome detachment, with the poles remaining stably associated with the cell cortex. Finally, we provide evidence that CaM is required for Asp's spindle function; however, it is completely dispensable for Asp's role in microcephaly suppression.