EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
A feedforward loop of NLRC5 (de)ubiquitination keeps IKK-NF-κB in check.
PMID 26620908
Abstract
Many receptors signal via adaptors to the IKK-NF-κB axis, transducing extracellular cues to transcriptional regulation. In this issue, Meng et al. (2015. J. Cell Biol. http://dx.doi.org/10.1083/jcb.201505091) reveal that the IKK regulator NLRC5 shapes NF-κB activity through a feedforward loop of NLRC5 ubiquitination and deubiquitination, highlighting a new pathway modulating IKK-NF-κB activity.