EMAIL THIS PAGE TO A FRIEND
Scientific reports
Optogenetics-enabled assessment of viral gene and cell therapy for restoration of cardiac excitability.
PMID 26621212
Abstract
Multiple cardiac pathologies are accompanied by loss of tissue excitability, which leads to a range of heart rhythm disorders (arrhythmias). In addition to electronic device therapy (i.e. implantable pacemakers and cardioverter/defibrillators), biological approaches have recently been explored to restore pacemaking ability and to correct conduction slowing in the heart by delivering excitatory ion channels or ion channel agonists. Using optogenetics as a tool to selectively interrogate only cells transduced to produce an exogenous excitatory ion current, we experimentally and computationally quantify the efficiency of such biological approaches in rescuing cardiac excitability as a function of the mode of application (viral gene delivery or cell delivery) and the geometry of the transduced region (focal or spatially-distributed). We demonstrate that for each configuration (delivery mode and spatial pattern), the optical energy needed to excite can be used to predict therapeutic efficiency of excitability restoration. Taken directly, these results can help guide optogenetic interventions for light-based control of cardiac excitation. More generally, our findings can help optimize gene therapy for restoration of cardiac excitability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3607
Cyanocobalamin, meets USP testing specifications
C63H88CoN14O14P
47869
Cyanocobalamin (B12), analytical standard
C63H88CoN14O14P
47829
D-(+) Glucose, analytical standard
C6H12O6
G7528
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
C6H12O6
G8270
D-(+)-Glucose, ≥99.5% (GC)
C6H12O6
G5767
D-(+)-Glucose, ACS reagent
C6H12O6
G7021
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
C6H12O6
G5400
D-(+)-Glucose, ≥99% (GC)
C6H12O6
G6152
D-(+)-Glucose, suitable for mouse embryo, ≥99.5% (GC)
C6H12O6
G5146
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
C6H12O6
G7520
D-(+)-Glucose, plant cell culture tested, BioReagent
C6H12O6
49139
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
16325
D-(+)-Glucose, meets analytical specification of Ph. Eur., BP, anhydrous
C6H12O6
608203
D-(+)-Glucose, 99.9 atom % 16O, 99.9 atom % 12C
C6H12O6
47249
D-(+)-Glucose, analytical standard
C6H12O6
D9434
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
C6H12O6
D8066
Dextrose, meets EP, BP, JP, USP testing specifications
C6H12O6
F8775
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
CH2O
F8923
Formaldehyde solution, meets USP testing specifications, contains 9.0-15% methanol as stabilizer
CH2O
252549
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
CH2O
F15587
Formaldehyde solution, 37 wt. % in H2O
CH2O
47608
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
CH2O
33220
Formaldehyde solution, puriss. p.a., ACS reagent, ≥36.5%
CH2O
15512
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
CH2O
47083-U
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, analytical standard
CH2O
606758
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
12CH2O
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
H4034
HEPES, BioPerformance Certified, ≥99.5% (titration), cell culture tested
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S