PloS one
A Zebrafish Model for Studies on Esophageal Epithelial Biology.
PMID 26630178
Abstract
Mammalian esophagus exhibits a remarkable change in epithelial structure during the transition from embryo to adult. However, the molecular mechanisms of esophageal epithelial development are not well understood. Zebrafish (Danio rerio), a common model organism for vertebrate development and gene function, has not previously been characterized as a model system for esophageal epithelial development. In this study, we characterized a piece of non-keratinized stratified squamous epithelium similar to human esophageal epithelium in the upper digestive tract of developing zebrafish. Under the microscope, this piece was detectable at 5dpf and became stratified at 7dpf. Expression of esophageal epithelial marker genes (Krt5, P63, Sox2 and Pax9) was detected by immunohistochemistry and in situ hybridization. Knockdown of P63, a gene known to be critical for esophageal epithelium, disrupted the development of this epithelium. With this model system, we found that Pax9 knockdown resulted in loss or disorganization of the squamous epithelium, as well as down-regulation of the differentiation markers Krt4 and Krt5. In summary, we characterized a region of stratified squamous epithelium in the zebrafish upper digestive tract which can be used for functional studies of candidate genes involved in esophageal epithelial biology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100432
Anti-CKAP4 (ab1) antibody produced in rabbit, affinity isolated antibody
HPA000278 Anti-CKAP4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000792
Anti-CKAP4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701800
Anti-sox2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
S9072 Anti-SOX2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
AV38232
Anti-SOX2 antibody produced in rabbit, IgG fraction of antiserum
SAB3500363
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104660
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB4300669
Anti-SOX2 antibody produced in rabbit, affinity isolated antibody
SAB2701973
Anti-SOX2 antibody produced in rabbit
SAB2701974
Anti-SOX2 antibody produced in rabbit
SAB2103958
Anti-SOX2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
S1451
Monoclonal Anti-Sox2 antibody produced in mouse, ~1.0 mg/mL, clone SOX2-6, purified immunoglobulin, buffered aqueous solution
SAB5300177
Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10, ascites fluid
SAB5300369 Monoclonal Anti-SOX2 antibody produced in mouse, clone 10F10C9, ascites fluid