EMAIL THIS PAGE TO A FRIEND
Dental update
A PREP Panel, Practice-Based, Evaluation of the Handling of the Kerr Demi-Ultra Light Curing Unit.
PMID 26630865
Abstract
This paper describes the handling evaluation (by a group of practice-based researchers, the PREP Panel) of a recently introduced Light Curing Unit (LCU), the Kerr Demi-Ultra, which possesses a number of novel features such as its ultracapacitor power source, and the Light Emitting Diodes (LEDs) which provide the light output being placed close to the tip of the light guide. CPD/CLINICAL RELEVANCE: Testing of new devices and materials with respect to their handling is of importance, given that an easy to handle device should produce better clinical results than one which is difficult to use.