EMAIL THIS PAGE TO A FRIEND
BMC bioinformatics
Accurate prediction of nuclear receptors with conjoint triad feature.
PMID 26630876
Abstract
Nuclear receptors (NRs) form a large family of ligand-inducible transcription factors that regulate gene expressions involved in numerous physiological phenomena, such as embryogenesis, homeostasis, cell growth and death. These nuclear receptors-related pathways are important targets of marketed drugs. Therefore, the design of a reliable computational model for predicting NRs from amino acid sequence has now been a significant biomedical problem. Conjoint triad feature (CTF) mainly considers neighbor relationships in protein sequences by encoding each protein sequence using the triad (continuous three amino acids) frequency distribution extracted from a 7-letter reduced alphabet. In addition, chaos game representation (CGR) can investigate the patterns hidden in protein sequences and visually reveal previously unknown structure. In this paper, three methods, CTF, CGR, amino acid composition (AAC), are applied to formulate the protein samples. By considering different combinations of three methods, we study seven groups of features, and each group is evaluated by the 10-fold cross-validation test. Meanwhile, a new non-redundant dataset containing 474 NR sequences and 500 non-NR sequences is built based on the latest NucleaRDB database. Comparing the results of numerical experiments, the group of combined features with CTF and AAC gets the best result with the accuracy of 96.30% for identifying NRs from non-NRs. Moreover, if it is classified as a NR, it will be further put into the second level, which will classify a NR into one of the eight main subfamilies. At the second level, the group of combined features with CTF and AAC also gets the best accuracy of 94.73%. Subsequently, the proposed predictor is compared with two existing methods, and the comparisons show that the accuracies of two levels significantly increase to 98.79% (NR-2L: 92.56 %; iNR-PhysChem: 98.18%; the first level) and 93.71% (NR-2L: 88.68%; iNR-PhysChem: 92.45%; the second level) with the introduction of our CTF-based method. Finally, each component of CTF features is analyzed via the statistical significant test, and a simplified model only with the resulting top-50 significant features achieves accuracy of 95.28%. The experimental results demonstrate that our CTF-based method is an effective way for predicting nuclear receptor proteins. Furthermore, the top-50 significant features obtained from the statistical significant test are considered as the "intrinsic features" in predicting NRs based on the analysis of relative importance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235202
4-Bromomethyl-7-methoxycoumarin, 97%
C11H9BrO3