EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
An Inducible, Large-Intestine-Specific Transgenic Mouse Model for Colitis and Colitis-Induced Colon Cancer Research.
PMID 26631394
Abstract
Animal models are an important tool to understand intestinal biology. Our laboratory previously generated C57BL/6-Tg(Car1-cre)5Flt transgenic mice (CAC) with large-intestine-specific Cre recombinase (Cre) expression as a model to study colon health. To expand the utility of the CAC mouse model by determining the impact of chemically induced colitis on CAC transgene expression. CAC mice were crossed to Rosa reporter mice (Rosa26R (flox/flox) ) with a lox-STOP-lox signal controlling β-galactosidase (βgal) expression and then further crossed with Apc(CKO/CKO) mice in some experiments to delete Apc alleles (Apc (Δ580) ). Initially, 8-week-old CAC(Tg/WT);Rosa26R (flox/WT) ;Apc (Δ580/WT) mice were treated with dextran sulfate sodium (DSS) in drinking water (5 days, 0, 0.65, 1.35, or 2.0 %). Colon tissue damage and βgal labeling were analyzed 10 day after stopping DSS. Next, 8-week-old CAC(Tg/WT);Rosa26R(flox/flox) mice were treated with 0 or 1.35 % DSS, and colonic βgal labeling was assessed at 30 day post-DSS treatment. Finally, 10-week-old CAC(Tg/WT);Apc (Δ580/WT) mice were treated with DSS (0 or 2 %) for 5 days and colonic tumors were analyzed at 20 weeks. CAC(Tg/WT);Rosa26R (flox/WT) ;Apc (Δ580/WT) mice had a DSS dose-dependent increase in colon epithelial damage that correlated with increased epithelial βgal labeling at 10 days (r (2) = 0.9, β = 0.75). The βgal labeling in CAC(Tg/WT);Rosa26R(flox/flox) mice colon remained high at 30 days, especially in the crypts of the healed ulcer. DSS also increased colon tumor incidence and multiplicity in CAC(Tg/WT);Apc (Δ580/WT) mice. DSS-mediated epithelial damage induces a persistent, Cre-mediated recombination of floxed alleles in CAC mice. This enables the examination of gene function in colon epithelium during experimental colitis and colitis-induced colon cancer.