EMAIL THIS PAGE TO A FRIEND
Scientific reports
Middle East respiratory syndrome coronavirus ORF4b protein inhibits type I interferon production through both cytoplasmic and nuclear targets.
PMID 26631542
Abstract
Middle East respiratory syndrome coronavirus (MERS-CoV) is a novel and highly pathogenic human coronavirus and has quickly spread to other countries in the Middle East, Europe, North Africa and Asia since 2012. Previous studies have shown that MERS-CoV ORF4b antagonizes the early antiviral alpha/beta interferon (IFN-α/β) response, which may significantly contribute to MERS-CoV pathogenesis; however, the underlying mechanism is poorly understood. Here, we found that ORF4b in the cytoplasm could specifically bind to TANK binding kinase 1 (TBK1) and IκB kinase epsilon (IKKε), suppress the molecular interaction between mitochondrial antiviral signaling protein (MAVS) and IKKε, and inhibit IFN regulatory factor 3 (IRF3) phosphorylation and subsequent IFN-β production. Further analysis showed that ORF4b could also inhibit IRF3 and IRF7-induced production of IFN-β, whereas deletion of the nuclear localization signal of ORF4b abrogated its ability to inhibit IRF3 and IRF7-induced production of IFN-β, but not IFN-β production induced by RIG-I, MDA5, MAVS, IKKε, and TBK-1, suggesting that ORF4b could inhibit the induction of IFN-β in both the cytoplasm and nucleus. Collectively, these results indicate that MERS-CoV ORF4b inhibits the induction of type I IFN through a direct interaction with IKKε/TBK1 in the cytoplasm, and also in the nucleus with unknown mechanism. Viruses have evolved multiple strategies to evade or thwart a host's antiviral responses. A novel human coronavirus (HCoV), Middle East respiratory syndrome coronavirus (MERS-CoV), is distinguished from other coronaviruses by its high pathogenicity and mortality. However, virulence determinants that distinguish MERS-CoV from other HCoVs have yet to be identified. MERS-CoV ORF4b antagonizes the early antiviral response, which may contribute to MERS-CoV pathogenesis. Here, we report the identification of the interferon (IFN) antagonism mechanism of MERS-CoV ORF4b. MERS-CoV ORF4b inhibits the production of type I IFN through a direct interaction with IKKε/TBK1 in the cytoplasm, and also in the nucleus with unknown mechanism. These findings provide a rationale for the novel pathogenesis of MERS-CoV as well as a basis for developing a candidate therapeutic against this virus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9542
DAPI, for nucleic acid staining
C16H15N5 · 2HCl
G3126
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
C5H10N2O3
G5792
L-Glutamine, PharmaGrade, Ajinomoto, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H10N2O3
G8540
L-Glutamine, meets USP testing specifications, cell culture tested, 99.0-101.0%, from non-animal source
C5H10N2O3
G6392
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
C5H10N2O3
49419
L-Glutamine, BioUltra, ≥99.5% (NT)
C5H10N2O3
W368401
L-Glutamine
C5H10N2O3
RES0926S-A7
Sodium chloride, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
ClNa
S7653
Sodium chloride, BioXtra, ≥99.5% (AT)
ClNa
S3014
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥98% (titration)
ClNa
S5886
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
ClNa
S1679
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
ClNa
S6150
Sodium chloride, tablet
ClNa
S6191
Sodium chloride, BioPerformance Certified, ≥99% (titration), Cell Culture Tested
ClNa
450006
Sodium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClNa
450014
Sodium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
ClNa
204439
Sodium chloride, 99.999% trace metals basis
ClNa
378860
Sodium chloride, random crystals, optical grade, 99.98% trace metals basis
ClNa
310166
Sodium chloride, reagent grade, ≥98%, +80 mesh particle size
ClNa
71376
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClNa
28-2270
Sodium chloride, JIS special grade, ≥99.5%
ClNa
S5150
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
ClNa
S6546
Sodium chloride solution, 5 M
ClNa
59222C
Sodium chloride solution, 5 M
ClNa
71386
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
ClNa
S0817
Sodium chloride solution, 0.85%
ClNa
16818
Sodium chloride solution, 2 M
ClNa
S8776
Sodium chloride solution, solution (0.9%), BioXtra, suitable for cell culture
ClNa
451940
Sodium chloride-35Cl, 99 atom % 35Cl
35ClNa
D6750
Sodium deoxycholate, ≥97% (titration)
C24H39NaO4