EMAIL THIS PAGE TO A FRIEND
Scientific reports
microRNA targeting of the P2X7 purinoceptor opposes a contralateral epileptogenic focus in the hippocampus.
PMID 26631939
Abstract
The ATP-gated ionotropic P2X7 receptor (P2X7R) modulates glial activation, cytokine production and neurotransmitter release following brain injury. Levels of the P2X7R are increased in experimental and human epilepsy but the mechanisms controlling P2X7R expression remain poorly understood. Here we investigated P2X7R responses after focal-onset status epilepticus in mice, comparing changes in the damaged, ipsilateral hippocampus to the spared, contralateral hippocampus. P2X7R-gated inward currents were suppressed in the contralateral hippocampus and P2rx7 mRNA was selectively uploaded into the RNA-induced silencing complex (RISC), suggesting microRNA targeting. Analysis of RISC-loaded microRNAs using a high-throughput platform, as well as functional assays, suggested the P2X7R is a target of microRNA-22. Inhibition of microRNA-22 increased P2X7R expression and cytokine levels in the contralateral hippocampus after status epilepticus and resulted in more frequent spontaneous seizures in mice. The major pro-inflammatory and hyperexcitability effects of microRNA-22 silencing were prevented in P2rx7(-/-) mice or by treatment with a specific P2X7R antagonist. Finally, in vivo injection of microRNA-22 mimics transiently suppressed spontaneous seizures in mice. The present study supports a role for post-transcriptional regulation of the P2X7R and suggests therapeutic targeting of microRNA-22 may prevent inflammation and development of a secondary epileptogenic focus in the brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8273 Anti-α-Amylase antibody produced in rabbit, fractionated antiserum, lyophilized powder
SAB1400010
Anti-AMY2A antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1410556
Anti-AMY2A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA039928
Anti-BLOC1S6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1303264
ANTI-EIF2C2 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV33799
Anti-EPB42 antibody produced in rabbit, affinity isolated antibody
HPA040261
Anti-EPB42 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
G4546 Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA044572
Anti-NPY antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500742
Anti-Pallidin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400488
Anti-PLDN antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2101826
Anti-PLDN antibody produced in rabbit, affinity isolated antibody
SAB2105000
Anti-PLDN, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA030790
Anti-RBFOX3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4200085
Monoclonal Anti-AGO2 antibody produced in rat, ~1.5 mg/mL, clone 11A9, purified immunoglobulin
WH0027161M1
Monoclonal Anti-EIF2C2 antibody produced in mouse, clone 2E12-1C9, purified immunoglobulin, buffered aqueous solution
SAB1405034
Monoclonal Anti-EIF2C2 antibody produced in mouse, clone 4D1-1D1, purified immunoglobulin, buffered aqueous solution
SAB1402184
Monoclonal Anti-EPB42, (C-terminal) antibody produced in mouse, clone 2G12, purified immunoglobulin, buffered aqueous solution
SAB1405011
Monoclonal Anti-PLDN antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution
SAB1402562
Monoclonal Anti-PLDN, (N-terminal) antibody produced in mouse, clone 2H8, purified immunoglobulin, buffered aqueous solution
S5768
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid