EMAIL THIS PAGE TO A FRIEND
FEBS letters
EZH2 phosphorylation regulates Tat-induced HIV-1 transactivation via ROS/Akt signaling pathway.
PMID 26632198
Abstract
EZH2 plays a major role in HIV-1 latency, however, the molecular linkage between Tat-induced HIV-1 transactivation and EZH2 activity is not fully understood. It was shown Tat induced HIV-1 transactivation through inhibiting EZH2 activity. Tat decreased the levels of H3K27me3 and EZH2 occupy at the long terminal repeat (LTR) of HIV-1. We further showed for the first time that transfected with Tat construct resulted in an increase in phosphorylated EZH2 (p-EZH2), mediated by active Akt. ROS/Akt-dependent p-EZH2 was correlated with Tat-induced transactivation. Our study reveals that novel mechanisms allow Tat-induced HIV-1 transactivation by ROS/Akt-dependent downregulating the EZH2 epigenetic silencing machinery.