EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
A GC-MS Based Metabonomics Study of Rheumatoid Arthritis and the Interventional Effects of the Simiaowan in Rats.
PMID 26633333
Abstract
Simiaowan (SMW) is a famous Chinese prescription widely used in clinical treatment of rheumatoid arthritis (RA). The aim of the present study is to determine novel biomarkers to increase the current understanding of RA mechanisms, as well as the underlying therapeutic mechanism of SMW, in RA-model rats. Plasma extracts from control, RA model, and SMW-treated rats were analyzed by gas chromatography coupled with mass spectrometry (GC-MS). An orthogonal partial least-square discriminant analysis (OPLS-DA) model was created to detect metabolites that were expressed in significantly different amounts between the RA model and the control rats and investigate the therapeutic effect of SMW. Metabonomics may prove to be a valuable tool for determining the efficacy of complex traditional prescriptions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M50400
N-Methylhydroxylamine hydrochloride, 98%
CH5NO · HCl