EMAIL THIS PAGE TO A FRIEND
Journal of biochemistry
Effects of the N terminus of mouse DNA polymerase κ on the bypass of a guanine-benzo[a]pyrenyl adduct.
PMID 26634445
Abstract
DNA polymerase κ (Polκ), one of the typical member of the Y-family DNA polymerases, has been demonstrated to bypass the 10S(+)-trans-anti-benzo[a]pyrene diol epoxide-N(2)-deoxyguanine adducts (BPDE-dG) efficiently and accurately. A large structural gap between the core and little finger as well as an N-clasp domain are essential to its unique translesion capability. However, whether the extreme N-terminus of Polκ is required for its activity is unclear. In this work, we constructed two mouse Polκ deletions, which have either a catalytic core (mPolκ1-516) or a core without the first 21-residues (mPolκ22-516), and tested their activities in the replication of normal and BPDE-DNA. These two Polκ deletions are nearly as efficient as the full length protein (Polκ1-852) in normal DNA synthesis. However, steady-state kinetics reveals a significant reduction in efficiency of dCTP incorporation opposite the lesion by Polκ22-516, along with increased frequencies for misinsertion compared with Polκ1-852 The next nucleotide insertion opposite the template C immediately following the BPDE-dG was also examined, and the bypass differences induced by deletions were highlighted in both insertion and extension step. We conclude that the extreme N-terminal part of Polκ is required for the processivity and fidelity of Polκ during translesion synthesis of BPDE-dG lesions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0662
6-Carboxyfluorescein, ~97% (HPLC)
C21H12O7