EMAIL THIS PAGE TO A FRIEND
Environmental health : a global access science source
Elevated concentrations of serum matrix metalloproteinase-2 and -9 and their associations with circulating markers of cardiovascular diseases in chronic arsenic-exposed individuals.
PMID 26637202
Abstract
Cardiovascular diseases (CVDs) and cancers are the major causes of chronic arsenic exposure-related morbidity and mortality. Matrix metalloproteinase-2 (MMP-2) and -9 (MMP-9) are deeply involved in the pathogenesis of CVDs and cancers. This study has been designed to evaluate the interactions of arsenic exposure with serum MMP-2 and MMP-9 concentrations especially in relation to the circulating biomarkers of CVDs. A total of 373 human subjects, 265 from arsenic-endemic and 108 from non-endemic areas in Bangladesh were recruited for this study. Arsenic concentrations in the specimens were measured by inductively coupled plasma mass spectroscopy (ICP-MS) and serum MMPs were quantified by immunoassay kits. Serum MMP-2 and MMP-9 concentrations in arsenic-endemic population were significantly (p < 0.001) higher than those in non-endemic population. Both MMPs showed significant positive interactions with drinking water (r s  = 0.208, p < 0.001 for MMP-2; r s  = 0.163, p < 0.01 for MMP-9), hair (r s  = 0.163, p < 0.01 for MMP-2; r s  = 0.173, p < 0.01 for MMP-9) and nail (r s  = 0.160, p < 0.01 for MMP-2; r s  = 0.182, p < 0.001 for MMP-9) arsenic of the study subjects. MMP-2 concentrations were 1.02, 1.03 and 1.05 times, and MMP-9 concentrations were 1.03, 1.06 and 1.07 times greater for 1 unit increase in log-transformed water, hair and nail arsenic concentrations, respectively, after adjusting for covariates (age, sex, BMI, smoking habit and hypertension). Furthermore, both MMPs were increased dose-dependently when the study subjects were split into three (≤10, 10.1-50 and > 50xa0μg/L) groups based on the regulatory upper limit of water arsenic concentration set by WHO and Bangladesh Government. MMPs were also found to be significantly (p < 0.05) associated with each other. Finally, the concentrations of both MMPs were correlated with several circulating markers related to CVDs. This study showed the significant positive associations and dose-response relationships of arsenic exposure with serum MMP-2 and MMP-9 concentrations. This study also showed the interactions of MMP-2 and MMP-9 concentrations with the circulating markers of CVDs suggesting the MMP-2 and MMP-9 -mediated mechanism of arsenic-induced CVDs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
202657
Arsenic, 99.999% trace metals basis
As
267961
Arsenic, powder, ≥99.997% trace metals basis
As
CKOZFN1039 CompoZr® Knockout ZFN Kit, human MMP2 (NM_004530)
CKOZFN1040 CompoZr® Knockout ZFN Kit, human MMP9 (NM_004994)
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
693642
Yttrium, foil, thickness 1 mm, 99.8% trace rare earth metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y
GF91377478
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Y
GF87149481
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Y
GF03037148
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Y
GF55118257
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Y
GF33100298
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Y
GF75315719
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Y
GF61507740
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Y
GF40879008
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Y
GF12674617
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Y
GF84327037
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Y
GF68048860
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Y
GF68472692
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Y
GF35512725
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Y
GF95617020
Yttrium, wire reel, 0.05m, diameter 1.0mm, square section, 99.9%
Y
GF02763963
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Y
GF39091158
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Y
GF79949255
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Y