EMAIL THIS PAGE TO A FRIEND
Lipids in health and disease
DDX5/p68 RNA helicase expression is essential for initiating adipogenesis.
PMID 26637310
Abstract
DDX5/p68 RNA helicase is a member of the DEAD (Asp-Glu-Ala-Asp) box proteins. Apart from RNA unwinding, DDX5 is an important transcriptional factor and co-activator in cell proliferation and differentiation. Here, we have characterised the role of DDX5 in adipogenesis in 3T3-L1 cells using siRNA mediated suppression. Transient inhibition of Ddx5 mRNA expression at the start of adipogenesis impairs the differentiation programme even when DDX5 expression is restored later in adipogenesis. However transient suppression of Ddx5 at the later stages of adipogenesis do not impair adipogenesis or triglyceride accumulation suggesting Ddx5 expression is dispensable in a mature adipocyte. These results implicate DDX5 as a crucial factor involved in the complex transcriptional cascade of events that regulate adipogenesis and essential to the initiation of adipogenesis.