EMAIL THIS PAGE TO A FRIEND
Scientific reports
The Dynamics and Regulatory Mechanism of Pronuclear H3k9me2 Asymmetry in Mouse Zygotes.
PMID 26639638
Abstract
H3K9 methylation is an important histone modification that is correlated with gene transcription repression. The asymmetric H3K9 dimethylation (H3K9me2) pattern between paternal and maternal genomes is generated soon after fertilization. In the present study, we carefully determined the dynamics of H3K9me2 changes in mouse zygotes, and investigated the regulatory mechanisms. The results indicated that histone methyltransferase G9a, but not GLP, was involved in the regulation of asymmetric H3K9me2, and G9a was the methyltransferase that induced the appearance of H3K9me2 in the male pronucleus of the zygote treated with cycloheximide. We found that there were two distinct mechanisms that regulate H3K9me2 in the male pronucleus. Before 8 h of in vitro fertilization (IVF), a mechanism exists that inhibits the association of G9a with the H3K9 sites. After 10 h of IVF the inhibition of G9a activity depends on yet unknown novel protein(s) synthesis. The two mechanisms of transfer take place between 8-10 h of IVF, and the novel protein failed to inhibit G9a activity in time, resulting in the appearance of a low level de novo H3K9me2 in the male pronucleus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2700645
Anti-Ehmt2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100657
Anti-EHMT2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA050550
Anti-EHMT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW22763 Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB3500247
Anti-GAPDH antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405848
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701826
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701825
Anti-gapdh antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
G9545
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2103104
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB4300645
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody
HPA040067
Anti-GAPDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2500450
Anti-GAPDH, C-Terminal antibody produced in goat, affinity isolated antibody, buffered aqueous solution
PLA0302
Goat anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
G8795
Monoclonal Anti-GAPDH antibody produced in mouse, ~1 mg/mL, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution
WH0002597M1
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
SAB1403850
Monoclonal Anti-GAPDH, (C-terminal) antibody produced in mouse, clone 1G5, purified immunoglobulin, buffered aqueous solution
PLA0125
Rabbit anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.