EMAIL THIS PAGE TO A FRIEND
Scientific reports
Developmental exposure to Ethinylestradiol affects transgenerationally sexual behavior and neuroendocrine networks in male mice.
PMID 26640081
Abstract
Reproductive behavior and physiology in adulthood are controlled by hypothalamic sexually dimorphic neuronal networks which are organized under hormonal control during development. These organizing effects may be disturbed by endocrine disrupting chemicals (EDCs). To determine whether developmental exposure to Ethinylestradiol (EE2) may alter reproductive parameters in adult male mice and their progeny, Swiss mice (F1 generation) were exposed from prenatal to peripubertal periods to EE2 (0.1-1 μg/kg/d). Sexual behavior and reproductive physiology were evaluated on F1 males and their F2, F3 and F4 progeny. EE2-exposed F1 males and their F2 to F4 progeny exhibited EE2 dose-dependent increased sexual behavior, with reduced latencies of first mount and intromission, and higher frequencies of intromissions with a receptive female. The EE2 1 μg/kg/d exposed animals and their progeny had more calbindin immunoreactive cells in the medial preoptic area, known to be involved in the control of male sexual behavior in rodents. Despite neuroanatomical modifications in the Gonadotropin-Releasing Hormone neuron population of F1 males exposed to both doses of EE2, no major deleterious effects on reproductive physiology were detected. Therefore EE2 exposure during development may induce a hypermasculinization of the brain, illustrating how widespread exposure of animals and humans to EDCs can impact health and behaviors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2502122
Anti-CALB1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA023099
Anti-CALB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408554
Anti-CALB1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
2858
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
C2H6O
48075
Ethanol solution, certified reference material, 2000 μg/mL in methanol
C2H6O
E2385
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
C2H6O
792780
Ethyl alcohol, Pure, 200 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792799
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
459836
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
C2H6O
792802
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
C2H6O
792829
Ethyl alcohol, Pure, 140 proof, meets water USP testing specifications, Excise Tax-free, Permit for use required
C2H6O
792810
Ethyl alcohol, Pure, 140 proof, Excise Tax-free, Permit for use required
C2H6O
493538
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
C2H6O
493511
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
C2H6O
P0130
Progesterone, ≥99%
C21H30O2
P6149
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
C21H30O2
P8783
Progesterone, powder, BioReagent, suitable for cell culture
C21H30O2
P3972
Progesterone, meets USP testing specifications
C21H30O2
X1126
Xylazine, ≥99%
C12H16N2S