EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Single-cell microinjection assay indicates that 7-hydroxycoumarin induces rapid activation of caspase-3 in A549 cancer cells.
PMID 26640551
Abstract
Coumarins have attracted intense interest in recent years due to their apoptogenic effects. The aim of the present study was to determine whether 7-hydroxycoumarin (7-HC) induces changes in caspase-3 (C-3) activity in A549 human lung carcinoma cells. A range of analytical techniques, including colorimetric and fluorometric assays, western blotting, single-cell microinjection, fluorescence microscopy and image analysis were conducted to elucidate the effects of 7-HC. A 24-h exposure to 1.85 mM 7-HC induced a 65% increase in C-3 activity, and a notable conversion of procaspase-3 to C-3, in addition to poly(ADP-ribose)polymerase cleavage. Furthermore, morphological changes associated with apoptosis were observed. Exposure of the cells to 7-HC for 3 or 6 h increased calcium conductance by 27%. By performing the single-cell microinjection of a specific fluorescent substrate of C-3 into previously 7-HC-exposed cells, a typical enzymatic kinetic profile of C-3 activation was identified a number of hours prior to the morphological and biochemical changes associated with apoptosis being observed. These results suggest that the rapid in vivo activation of C-3 is induced by 7-HC, the most relevant biotransformation product of coumarin in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2559
N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide, ≥97% (HPLC), powder
C26H34N6O13