EMAIL THIS PAGE TO A FRIEND
Scientific reports
Induction of autophagy improves embryo viability in cloned mouse embryos.
PMID 26643778
Abstract
Autophagy is an essential cellular mechanism that degrades cytoplasmic proteins and organelles to recycle their components. Moreover, autophagy is essential for preimplantation development in mammals. Here we show that autophagy is also important for reprogramming in somatic cell nuclear transfer (SCNT). Our data indicate that unlike fertilized oocytes, autophagy is not triggered in SCNT embryos during 6 hours of activation. Mechanistically, the inhibited autophagic induction during SCNT activation is due to the cytochalasin B (CB) caused depolymerization of actin filaments. In this study, we induced autophagy during SCNT activation by rapamycin and pp242, which could restore the expected level of autophagy and significantly enhance the development of SCNT embryos to the blastocyst stage when compared with the control (68.5% and 68.7% vs. 41.5%, P < 0.05). Furthermore, the treatment of rapamycin and pp242 accelerates active DNA demethylation indicated by the conversion of 5 mC to 5 hmC, and treatment of rapamycin improves degradation of maternal mRNA as well. Thus, our findings reveal that autophagy is important for development of SCNT embryos and inhibited autophagic induction during SCNT activation might be one of the serious causes of low efficiency of SCNT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D27802
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
C6H12N2
290734
Dabco® 33-LV
C6H12N2
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
H4034
HEPES, BioPerformance Certified, ≥99.5% (titration), cell culture tested
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
RES6003H-B7
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
H7523
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
C8H18N2O4S
H6147
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
C8H18N2O4S
54457
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
C8H18N2O4S
83264
HEPES buffer solution, 1 M in H2O
C8H18N2O4S
H4272
Hyaluronidase from bovine testes, Type IV-S, powder, mouse embryo tested, 750-3000 units/mg solid
P6148
Paraformaldehyde, reagent grade, crystalline
CH2O ]n
PVP360
Polyvinylpyrrolidone, average mol wt 360,000
R8781 Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
C51H79NO13
553468
Tris(tert-butoxy)silanol, 99.999%
C12H28O4Si
697281
Tris(tert-butoxy)silanol, packaged for use in deposition systems
C12H28O4Si