EMAIL THIS PAGE TO A FRIEND
Medicine, science, and the law
Serum strontium estimation as a medico-legal diagnostic indicator of drowning.
PMID 2664410
Abstract
In order to corroborate the medico-legal diagnosis of vital submersion we reviewed the appropriate literature and tested Icard's initial hypothesis, already expressed in 1932, that Strontium might be a good indicator of sea-water drowning. Therefore, we examined all the bodies found either in fresh or in sea-water, and also non-drowned control cases. Strontium concentrations in both cardiac cavities and in a peripheral blood-vessel were determined by flameless atomic absorption spectrophotometry. The statistical evaluation of the results is hereby presented. We come to the conclusion that this determination can be a valuable additional proof for sea-water drowning diagnosis, whilst freshwater drowning would be more difficult to assess. Further investigations are being undertaken.