EMAIL THIS PAGE TO A FRIEND
Scientific reports
Regulation of Shootin1 Gene Expression Involves NGF-induced Alternative Splicing during Neuronal Differentiation of PC12 Cells.
PMID 26648138
Abstract
Shootin1 is a protein involved in neuronal polarization, and has been shown to be a key molecule for the positive/negative feedback loop for axon induction required during neuronal symmetry breaking. To better understand the molecular basis of shootin1 dynamics, we analysed the regulatory pathways and the expressional status of shootin1 gene during NGF-induced neuronal differentiation. We demonstrated that the isoform-1 and isoform-2 of shootin1 is differentially expressed during neuronal differentiation. By blocking individual downstream pathways of NGF signalling, we found that PI3K/Akt pathway plays a major role in the expression of shootin1 isoform-2. Western blot and RT-PCR results showed that the isoform-1 of shootin1 is constitutively expressed, while the isoform-2 is expressed in a manner that is strictly dependent on NGF-stimulation. Isoform-specific RT-PCR results demonstrated that the differential expression of the isoform-1 and isoform-2 of shootin1 is a consequence of alternative splicing of shootin1 pre-mRNA, in response to NGF-signalling. Collectively these findings provide the first information on the molecular mechanisms regulating the expression of shootin1 gene and represent the first example of NGF-induced alternative splicing process that has a regulatory role in neuritogenesis.