EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
SNAIL transcription factor increases the motility and invasive capacity of prostate cancer cells.
PMID 26648419
Abstract
The incidence and mortality rates of prostate cancer (PCa) are increasing, and PCa is almost the second‑leading cause of cancer‑associated mortality in men. During tumor progression, epithelial cells decrease the number of adhesion molecules, change their polarity and position, rearrange their cytoskeleton and increase their migratory and invasive capacities. These changes are known under the concept of epithelial‑mesenchymal transition (EMT). EMT is characterized by an upregulation of certain transcription factors, including SNAIL1, which represses genes that are characteristic of an epithelial phenotype, including E‑cadherin, and indirectly increase the expression levels of genes, which are associated with the mesenchymal phenotype. It has been suggested that the transcription factor, SNAIL1, decreases the proliferation and increases the migratory and invasive capacities of PCa cell lines. The present study was performed using LNCaP and PC3 cell lines, in which the expression levels of SNAIL1 were increased or silenced through the use of lentiviral vectors. The expression levels of EMT markers were quantified using reverse transcription‑quantitative polymerase chain reaction and western blot analysis. In addition, cell survival was analyzed using an MTS assay; cell proliferation was examined using an antibody targeting Ki‑67; migration on plates with 8 µm pores to allow the passage of cells; and invasiveness was analyzed using a membrane chamber covered in dried basement membrane matrix solution. The levels of apoptosis were determined using a Caspase 3/7 assay containing a substrate modified by caspases 3 and 7. The results demonstrated that the overexpression and silencing of SNAIL1 decreased cell proliferation and survival. However, the overexpression of SNAIL1 decreased apoptosis, compared with cells with the SNAIL1‑silenced cells, in which cell apoptosis increased. The migration and invasive capacities increased in the cells overexpressing SNAIL1, and decreased when SNAIL1 was silenced. In conclusion, PCa cells overexpressing SNAIL1 exhibited characteristics of an EMT phenotype, whereas the silencing of the SNAIL1 transcriptional repressor promoted an epithelial‑like phenotype, with decreased migration and invasion, characteristic of mesenchymal cells.