EMAIL THIS PAGE TO A FRIEND
Stem cells and development
The Effect of Simulated Microgravity on Differentiation of Porcine Blood-Derived Vascular Stem Cells.
PMID 26649474
Abstract
The negative effects of space flight on cardiovascular health of astronauts have been demonstrated and documented over many years. Endothelial cells (ECs) play an important role in regulating weightlessness-induced cardiovascular dysfunction. This project seeks to study the effect of microgravity on the differentiation of endothelial progenitor cells (EPCs) into ECs and the downstream functions of the differentiated cells. Initial exposure of EPCs to microgravity indicated an inhibition of migratory, proliferative, and antithrombogenic capacity of the differentiated ECs. However, our results indicate a potential recovery of proliferative and antithrombogenic functions on prolonged exposure to microgravity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33972
3,5,6-Trichloro-2-pyridinol, PESTANAL®, analytical standard
C5H2Cl3NO