EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Ultra-Performance Liquid Chromatographic Determination of Manufacturing Intermediates and Subsidiary Colors in D&C Red No. 6, D&C Red No. 7, and Their Lakes.
PMID 26651589
Abstract
An ultra-performance LC (UPLC) method was developed to determine the manufacturing intermediates and subsidiary colors in the monosulfo monoazo color additives D&C Red No. 6 and D&C Red No. 7 and their lakes. This method is intended for use in batch certification of the color additives by the U. S. Food and Drug Administration to ensure that each lot meets published specifications for coloring drugs and cosmetics. The intermediates are 2-amino-5-methylbenzenesulfonic acid (PTMS) and 3-hydroxy-2-naphthalenecarboxylic acid (3-hydroxy-2-naphthoic acid). The subsidiary colors are 3-hydroxy-4-[(4-methylphenyl)azo]-2-naphthalenecarboxylic acid (unsulfonated subsidiary color) and 1-[(4-methylphenyl) azo]-2-naphthalenol (4-methyl Sudan I). The analytes were identified by comparing their UPLC retention times and UV-Vis absorption spectra with those of standards. Validation studies showed that calibration curves were linear (average R2=0.9994), and recoveries were 96-106%. Average LOD was 0.0014-0.0061% and average LOQ was 0.0047-0.020%. Results for RSD at the specification levels ranged from 0.67 to 5.79%. Survey analyses of 42 samples from 14 domestic and foreign manufacturers yielded results by the new UPLC method and a previously reported HPLC method that were consistent within experimental error. The new UPLC method provided increased sensitivity, faster analysis times, and improved separations compared to the HPLC method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0130
Oxalic acid dilithium salt
C2Li2O4