EMAIL THIS PAGE TO A FRIEND
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology
Candidate genes for individual recognition in Polistes fuscatus paper wasps.
PMID 26660069
Abstract
Few animals are known to individually recognize conspecifics, i.e. learn and recall unique individuals during subsequent encounters, and nearly all are social vertebrates. Remarkably, the social paper wasp Polistes fuscatus has recently been discovered to possess this ability, which is useful for remembering identities during competitive social interactions. We analyzed brain gene expression in staged encounters between pairs of individuals to explore potential mechanisms underlying wasps' ability to recall familiar individuals using real-time qRT-PCR. We identified four candidate genes (IP3K, IP3R, Nckx30C and Su(var)2-10) that were down-regulated in the presence of familiar individuals compared to single wasps and pairs of wasps meeting for the first time. These candidate genes are related to calcium signaling, therefore, we treated wasps with lithium chloride, a pharmacological agent that inhibits calcium signaling in neurons. This treatment decreased aggression in paper wasps, but did not affect expression of genes related to calcium signaling. The results suggest calcium signaling differences may be related to individual memory recall in wasps, and we present four promising candidate genes for future study. These data suggest genes associated with dominance behavior may be co-opted for individual recognition, but further work is needed to establish a causal association with the behavior.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8640
E-64d, protease inhibitor
C17H30N2O5